missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ One Block Analog Dry Block Heater HB1AL

Product Code. 15762699
Click to view available options
Description:
Dry Block Heater, 1 Block, HB1AL, EU
This item is not returnable. View return policy
This item is not returnable. View return policy

OHAUS™ One Block Analog Dry Block Heater processes more samples while delivering precise and reliable results.

  • High wattage, constant-temperature Dry Block Heaters are economical, versatile and compact
  • Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
  • Each of the five models accept separate interchangeable modular blocks (supplied separately) accommodating various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
  • Each block has a thermometer well for measuring block temperature
  • Anodised aluminum modular blocks provide superior temperature stability and heat transfer
  • Analogue controls
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance TUV
Controller Type Analog
Description Dry Block Heater, 1 Block, HB1AL, EU
Temperature Range (Metric) 5°C to 100°C (Low), 75°C to 150°C (High)
Type Dry Bath Block
Heat Up Time 45 min
Model HB1AL
No. of Blocks 1
Temperature Stability ±1°C
Temperature Uniformity ±0.1°C at 37°C
Uniformity ± 0.4°C at 37°C
Dimensions (L x W) 31.50 cm x 20.30 cm
Dimensions (L x W x H) 31.5 x 20.3 x 8.9 cm
Length (English) 12.40 in.
Length (Metric) 31.50 cm
Width (English) 8.00 in.
Width (Metric) 20.30 cm
Height (English) 3.50 in.
Height (Metric) 8.900 cm
Depth (English) 12.40 in.
Depth (Metric) 31.50 cm
Weight (Metric) 2.5 kg
Electrical Requirements 230V, 0.5A, 50/60Hz
Amperage 0.5A
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'