Learn More
Macherey-Nagel™ Octyl Phase Polygosil™ C8
Brand:  Macherey-Nagel™ 711300.10
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10621102
  • £178.00 / 10g
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Specifications
5 μm
Liquid Chromatography
Polygosil C8 Phases
60 Å
10 g