Learn More
Macherey-Nagel™ Octyl Phase Polygosil™ C8
Brand:  Macherey-Nagel™ 711320.10
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10216361
  • £150.00 / 10g
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Specifications
10 μm
Liquid Chromatography
Polygosil C8 Phases
60 Å
10 g