missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Nuclear Fast Red, 94%, pure
£38.50 - £78.70
Chemical Identifiers
CAS | 6409-77-4 |
---|---|
Molecular Formula | C14H11NNaO7S |
Molecular Weight (g/mol) | 360.292 |
MDL Number | MFCD00078493 |
InChI Key | KDEATZKCHKVXGA-UHFFFAOYSA-N |
Synonym | Calcium Red, C.I. 60760 |
PubChem CID | 131675027 |
IUPAC Name | 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium |
SMILES | [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10144351
|
Thermo Scientific Chemicals
211980010 |
1 g | Glass bottle |
£38.50
1g |
In Stock
Log in to see stock. |
||||
10711745
|
Thermo Scientific Chemicals
|
5 g | Glass bottle |
£78.70
5g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6409-77-4 | |
360.292 | |
KDEATZKCHKVXGA-UHFFFAOYSA-N | |
131675027 | |
[HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na] |
C14H11NNaO7S | |
MFCD00078493 | |
Calcium Red, C.I. 60760 | |
4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium |
Specifications
>300.0°C | |
92.0 | |
94% | |
MFCD00078493 | |
Solubility in water: 1g/L (25°C). | |
[HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na] | |
360.292 | |
357.26 | |
Pure |
6409-77-4 | |
100.0 | |
C14H11NNaO7S | |
Calcium Red, C.I. 60760 | |
KDEATZKCHKVXGA-UHFFFAOYSA-N | |
4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium | |
131675027 | |
94% | |
Nuclear Fast Red |
Safety and Handling
missing translation for 'einecsNumber' : 229-088-0
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Nuclear Fast Red
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only