missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPS 2390, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Group I mGlu antagonist; displays noncompetitive antagonist activity at both mGlu1 and mGlu5 receptors. Thought to act on a site separate from the glutamate binding pocket.
Specifications
Quantity | 10 mg |
Formula Weight | 307.39 |
Percent Purity | >99% |
Chemical Name or Material | NPS 2390 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'