Learn More
Invitrogen™ Novex™ TBE Gels, 6%
Novex™ TBE Gels 6% are designed to run on the XCell SureLock™ Mini-Cell and provide high-resolution analysis of restriction digests and PCR products.
£183.00 - £187.00
Specifications
Content And Storage | • Catalog number refers to a single box of 10 gels Store at 4°C. |
---|---|
For Use With (Equipment) | XCell SureLock Mini-Cell |
Gel Percentage | 6% |
Gel Type | TBE |
Product Type | TBE Gel |
Product Code | Brand | Description | Wells | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Wells | Price | Quantity & Availability | ||||
12085725
|
Invitrogen™
EC6261BOX |
Novex TBE Gels, 6%, 1-well | 1-well |
£187.00
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
12095725
|
Invitrogen™
EC6265BOX |
Novex TBE Gels, 6%, 10-well | 10-well |
£185.00
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
12005735
|
Invitrogen™
EC62652BOX |
Novex TBE Gels, 6%, 12-well | 12-well |
£183.00
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
12015735
|
Invitrogen™
EC62655BOX |
Novex TBE Gels, 6%, 15-well | 15-well |
£183.00
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
Description
Novex™ TBE Gels 6% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 65–250 bp.
Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis.
Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results
Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.
Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.
Order Info
Shipping Condition: Wet Ice
Specifications
• Catalog number refers to a single box of 10 gels Store at 4°C. |
|
6% | |
TBE Gel | |
10 Gels/Box | |
Wet Ice |
XCell SureLock Mini-Cell | |
TBE | |
65 to 250 bp | |
Double-stranded DNA (dsDNA) |