Learn More
Invitrogen™ RANTES Rat ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ KRC1031
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Rat RANTES ELISA quantitates Rt RANTES in human serum, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt RANTES. Principle of the method The Rat RANTES solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Recognizes natural and recombinant IP-10.Order Info
Shipping Condition: Room Temperature
Specifications
P50231 | |
<20 pg/mL | |
ELISA Kit | |
Rat | |
Colorimetric Microplate Reader | |
Rantes,Scya5 | |
5.4% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RANTES | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
39-2500 pg/mL | |
Biotin | |
Serum, Supernatant | |
Colorimetric Detection | |
81780 | |
4 hr. | |
3.3% | |
HRP | |
96 Tests | |
Serum, 10 μL; Supernatant, 100 μL | |
Rat | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.