Learn More
Invitrogen™ InVision™ His-Tag In-Gel Staining Kit
One ready-to-use solution that does not require mixing, dilution, or solution preparation
Brand: Invitrogen™ LC6033
Code : 50
Additional Details : Weight : 1.67000kg
Includes: InVision™ His-tag In-gel Stain is supplied as a 1X ready-to-use staining reagent (500mL). BenchMark™ His-tagged Protein Standard (125 μL) is also included in the kit.
Description
The InVision™ His-tag In-gel Staining Kit includes InVision™ His-tag In-gel Stain (also available separately), as well as BenchMark™ His-tagged Protein Standard. InVision™ His-tag In-gel Stain is a ready-to-use, fast, sensitive and highly specific stain for visualizing His-tagged fusion protein bands directly in a polyacrylamide gel following electrophoresis.
- Stain proteins fused to oligohistidine sequence
- Eliminate western blotting-detect His-tagged proteins directly in the gel
- Detect nanogram levels of protein bands with UV transilluminator, equipped with standard camera, or laser-based scanner
- Visualize results in under 3 hours
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Specifications
InVision™ His-tag In-gel Stain is supplied as a 1X ready-to-use staining reagent (500 mL). BenchMark™ His-tagged Protein Standard (125 μL) is also included in the kit. | |
Fluorescent | |
His Tag Proteins In-Gel Staining Kit | |
Proteins (His-Tagged) |
In-Gel Detection | |
InVision His-Tag Stain | |
InVision™ |
For Research Use Only. Not for use in diagnostic procedures.