Learn More
Invitrogen™ IL-12 p70 Mouse ELISA Kit
Quantifies natural and recombinant mouse IL-12 heterodimer and the free p40 unit in serum, buffered solution, or cell culture medium
Brand: Invitrogen™ KMC0121
Code : 50
Additional Details : Weight : 0.70000kg
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
IL-12 is a lymphokine.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
Specifications
7.8-500 pg/mL | |
Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. | |
Coated ELISA Kit | |
Cytokines & Receptors | |
Mouse | |
Microplate Reader | |
IL-12p35, Il-12a, Ll12a, p35, Il-12b, Il-12p40, Il12p40, p40 | |
3 hr | |
Interleukin-12 subunit alpha, Interleukin-12 subunit beta | |
50 μL | |
IL-12 p70 | |
P43431, P43432 |
<2 pg/mL | |
Colorimetric | |
ELISA Kit | |
Cell Culture Supernatant, Serum | |
Novex | |
interleukin 12A, interleukin 12b | |
IL12A, IL12B | |
HRP | |
IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit, CLMF p35, IL-12 subunit p35, IL-12B, Cytotoxic lymphocyte maturation factor 40 kDa subunit, CLMF p40, IL-12 subunit p40 | |
Wet Ice | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.