missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(Ii) Chloride Hexahydrate, ReagentPlus™ Honeywell™
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Quantity | 2 kg |
Linear Formula | NiCl2 · 6H2O |
UN Number | UN3288 |
Formula Weight | 237.69g/mol |
Grade | ReagentPlus™ |
Packaging | Poly Bottle |
Chemical Name or Material | Nickel(Ii) Chloride Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'