missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium wire, 0.41mm (0.016in) dia, Thermo Scientific Chemicals

£30.50

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
IUPAC Name chromium;nickel
SMILES [Cr].[Ni]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379847
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042196.G9
25 m
£30.50
25m
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

These alloys are typically used in resistance wire. They are also used in some dental restorations (fillings). It is commonly used in ceramic as an internal support. Nichrome wire is used for its ability to withstand the high temperatures that occur when clay work is fired in a kiln.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

11106-97-1
110.69
VNNRSPGTAMTISX-UHFFFAOYSA-N
11320920
[Cr].[Ni]
CrNi
MFCD00673110
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
chromium;nickel

Specifications

11106-97-1
CrNi
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
VNNRSPGTAMTISX-UHFFFAOYSA-N
chromium;nickel
11320920
≈1.09g/m
Nickel Chromium wire, 0.41mm (0.016 in.) dia.
Ni:Cr; 80:20 wt%
MFCD00673110
Insoluble in water.
[Cr].[Ni]
110.69
Odorless
Wire
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only