missing translation for 'onlineSavingsMsg'
Learn More

NEMF Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA549768

Additional Details : Weight : 0.01000kg

Product Code. 15693695

  • £403.00 / 100µL
Estimated Shipment: 20-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody detects endogenous levels of total NEMF protein.

NEMF Polyclonal antibody specifically detects NEMF in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

NEMF
Polyclonal
Unconjugated
NEMF
1500011I12Rik; 4933405E14Rik; Antigen NY-CO-1; Nemf; nuclear export mediator factor; nuclear export mediator factor Nemf; NY-CO-1; SDCCAG1; Serologically defined colon cancer antigen 1; serologically defined colon cancer antigen 1 homolog
Rabbit
Antigen affinity chromatography
RUO
100322884, 66244, 9147
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O60524, Q8CCP0
NEMF
Synthesized peptide derived from internal of human SDCG1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers