missing translation for 'onlineSavingsMsg'
Learn More

NECAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17906-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16817914

  • £130.00 / 20µL
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Involved in endocytosis.
Specifications

Specifications

NECAP1
Polyclonal
Unconjugated
NECAP1
NECAP 1, NECAP endocytosis associated 1, NECAP1
Rabbit
Antigen Affinity Chromatography
RUO
25977, 312694, 67602
-20°C
Liquid
Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P69682, Q8NC96, Q9CR95
Necap1
NECAP1 Fusion Protein Ag12283
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers