Learn More
Thermo Scientific™ National Preassembled Target DP™ Kits
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Brand:  Thermo Scientific™ C4000-188WR
 View more versions of this product
Product Code. 11981593
Description
Description
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
  • Packaged in convenient vial trays with clear covers or in polybags
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Amber Glass I-D; Red
Glass
PTFE/Silicone/PTFE
Yes
Red/White/Red
Amber
2 mL
9-425
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ National Preassembled Target DP™ Kits > Amber Glass I-D; Red