missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NADPH tetrasodium salt hydrate, 96%, extra pure, Thermo Scientific Chemicals
£502.00 - £1337.00
Chemical Identifiers
CAS | 2646-71-1 |
---|---|
Molecular Formula | C21H26N7Na4O17P3 |
Molecular Weight (g/mol) | 833.35 |
MDL Number | MFCD10567216 |
InChI Key | WYWWVJHQDVCHKF-NRTBITFTNA-J |
Synonym | nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form |
PubChem CID | 131673986 |
IUPAC Name | [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
SMILES | [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10337241
|
Thermo Scientific Acros
328742500 |
250 mg | Glass bottle |
£502.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10125503
|
Thermo Scientific Acros
328740010 |
1 g | Glass bottle |
£1337.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2646-71-1 | |
833.35 | |
WYWWVJHQDVCHKF-NRTBITFTNA-J | |
131673986 | |
[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
C21H26N7Na4O17P3 | |
MFCD10567216 | |
nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
Specifications
2646-71-1 | |
100.0 | |
C21H26N7Na4O17P3 | |
nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form | |
WYWWVJHQDVCHKF-NRTBITFTNA-J | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium | |
131673986 | |
96% | |
NADPH tetrasodium salt hydrate, approx. 98% |
97.5 | |
95% min. (HPLC) | |
MFCD10567216 | |
Solubility in water: soluble | |
[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
833.35 | |
833.35 | |
Extra Pure |
Safety and Handling
MOISTURE SENSITIVE
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 220-163-3
RUO – Research Use Only