missing translation for 'onlineSavingsMsg'
Learn More

N-n-Hexylaniline, 98%, Thermo Scientific™

Product Code. 15431818
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4746-32-1
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00462723
InChI Key OXHJCNSXYDSOFN-UHFFFAOYSA-N
Synonym benzenamine, n-hexyl, hexylaniline, hexyl-phenyl-amine, benzenamine,n-hexyl, n-hexylaniline, acmc-20ak47
PubChem CID 138340
IUPAC Name N-hexylaniline
SMILES CCCCCCNC1=CC=CC=C1

Specifications

Color Colorless to Brown
Density 0.899g/mL
Boiling Point 140°C to 145°C (15 mmHg)
Refractive Index 1.5230
Quantity 1 g
Formula Weight 177.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-n-Hexylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'