missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine, 97%, Thermo Scientific Chemicals

Product Code. 10772865
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-98-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007860
InChI Key BZORFPDSXLZWJF-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline
PubChem CID 7472
ChEBI CHEBI:15783
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(N)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 35.0°C
Color Brown
Density 1.0900g/mL
Boiling Point 262.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2C6H4N(CH3)2
Quantity 500 g
Merck Index 15, 3278
Specific Gravity 1.09
Solubility Information Solubility in water: 11g/L (20°C). Other solubilities: soluble in alcohol,chloroform,ether,benzene
Formula Weight 136.2
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N, N-Dimethyl-p-phenylenediamine, Practical
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Dimethyl-p-phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'