Learn More
N-Methylphenethylamine, 99%, Thermo Scientific Chemicals
£51.80 - £505.00
Chemical Identifiers
CAS | 589-08-2 |
---|---|
Molecular Formula | C9H13N |
Molecular Weight (g/mol) | 135.21 |
MDL Number | MFCD00008291 |
InChI Key | SASNBVQSOZSTPD-UHFFFAOYSA-N |
Synonym | n-methylphenethylamine, benzeneethanamine, n-methyl, methyl-phenethyl-amine, n-methyl-2-phenylethylamine, n-methyl-n-2-phenylethyl amine, 1-phenyl-2-methylaminoethane, phenethylamine, n-methyl, methyl 2-phenylethyl amine, 1-phenyl-2-methylamino-aethan, n-methyl-phenethylamine |
PubChem CID | 11503 |
IUPAC Name | N-methyl-2-phenylethanamine |
SMILES | CNCCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10367150
|
Thermo Scientific Acros
127450050 |
5 g | Glass bottle |
£51.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10572331
|
Thermo Scientific Acros
127450250 |
25 g | Glass bottle |
£182.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617294
|
Thermo Scientific Acros
127451000 |
100 g | Glass Bottle |
£505.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
589-08-2 | |
135.21 | |
SASNBVQSOZSTPD-UHFFFAOYSA-N | |
11503 | |
CNCCC1=CC=CC=C1 |
C9H13N | |
MFCD00008291 | |
n-methylphenethylamine, benzeneethanamine, n-methyl, methyl-phenethyl-amine, n-methyl-2-phenylethylamine, n-methyl-n-2-phenylethyl amine, 1-phenyl-2-methylaminoethane, phenethylamine, n-methyl, methyl 2-phenylethyl amine, 1-phenyl-2-methylamino-aethan, n-methyl-phenethylamine | |
N-methyl-2-phenylethanamine |
Specifications
589-08-2 | |
100.0 | |
0.9300g/mL | |
73°C | |
98.5% min. (GC) | |
C6H5CH2CH2NHCH3 | |
MFCD00008291 | |
0.93 | |
Solubility in water: slightly soluble. Other solubilities: slightly soluble in alcohol and ether | |
CNCCC1=CC=CC=C1 | |
135.21 | |
135.21 | |
Liquid |
98.5 | |
Colorless to Yellow | |
203.0°C | |
Authentic | |
C9H13N | |
1.5150 to 1.5170 | |
12, 1097 | |
n-methylphenethylamine, benzeneethanamine, n-methyl, methyl-phenethyl-amine, n-methyl-2-phenylethylamine, n-methyl-n-2-phenylethyl amine, 1-phenyl-2-methylaminoethane, phenethylamine, n-methyl, methyl 2-phenylethyl amine, 1-phenyl-2-methylamino-aethan, n-methyl-phenethylamine | |
SASNBVQSOZSTPD-UHFFFAOYSA-N | |
N-methyl-2-phenylethanamine | |
11503 | |
99% | |
N-Methylphenethylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-632-3
- N-Methylphenethylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only