Learn More
N-Methylformamide, 99%, Thermo Scientific Chemicals
Description
N-Methylformamide is used to prepare methyl isocyanate. It is used as an extraction solvent for aromatic hydrocarbons and acts as a ligand in coordination chemistry. It is employed as a solvent in oil refineries and in aluminum electrolytic capacitors. Further, it is used as a reagent in organic synthesis. In addition to this, it is used as an investigational anticancer drug and an intermediate in the production of some pharmaceutical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Methylformamide is used to prepare methyl isocyanate. It is used as an extraction solvent for aromatic hydrocarbons and acts as a ligand in coordination chemistry. It is employed as a solvent in oil refineries and in aluminum electrolytic capacitors. Further, it is used as a reagent in organic synthesis. In addition to this, it is used as an investigational anticancer drug and an intermediate in the production of some pharmaceutical compounds.
Solubility
Miscible with water, acetone and ethanol. Immiscible with ether.
Notes
Incompatible with strong oxidizing agents, acids, bases and acid chlorides.
Specifications
Melting Point | -4°C |
Density | 1.018 |
Boiling Point | 181°C to 185°C |
Flash Point | 98°C (208°F) |
Linear Formula | HCONHCH3 |
Refractive Index | 1.432 |
Quantity | 50 g |
Beilstein | 1098352 |
Merck Index | 14,6076 |
Solubility Information | Miscible with water,acetone and ethanol. Immiscible with ether. |
Show More |
RUO – Research Use Only