missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylphthalimide, 98%, Thermo Scientific Chemicals

Product Code. 11438636
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5022-29-7
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00014583
InChI Key JZDSOQSUCWVBMV-UHFFFAOYSA-N
Synonym n-ethylphthalimide, 2-ethylisoindoline-1,3-dione, phthalimide, n-ethyl, 1h-isoindole-1,3 2h-dione, 2-ethyl, 2-ethyl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, n-ethyl, 2-ethylbenzo c azoline-1,3-dione, acmc-1az90, 1h-isoindole-1, n-ethyl
PubChem CID 21120
IUPAC Name 2-ethylisoindole-1,3-dione
SMILES CCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 74°C to 78°C
Quantity 25 g
Beilstein 132094
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material N-Ethylphthalimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'