missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-trans-4-hydroxy-L-proline, 98%, Thermo Scientific Chemicals

£114.57 - £321.00

Chemical Identifiers

CAS 33996-33-7
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
MDL Number MFCD00037339
InChI Key BAPRUDZDYCKSOQ-RITPCOANSA-N
Synonym oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum
PubChem CID 65784
IUPAC Name (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CC(CC1C(=O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11420218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23767.09
10 g
£114.57
10g
Please to purchase this item. Need a web account? Register with us today!
11430218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23767.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33996-33-7
173.168
BAPRUDZDYCKSOQ-RITPCOANSA-N
65784
CC(=O)N1CC(CC1C(=O)O)O
C7H11NO4
MFCD00037339
oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum
(2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid

Specifications

131°C to 133°C
C7H11NO4
84043
oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum
CC(=O)N1CC(CC1C(=O)O)O
173.168
173.17
Crystalline Powder
33996-33-7
MFCD00037339
Glass Bottle
BAPRUDZDYCKSOQ-RITPCOANSA-N
(2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
65784
99%
N-Acetyl-trans-4-hydroxy-L-proline
Videos
Safety and Handling

Safety and Handling

P280-P305+P351+P338-P310

H318

missing translation for 'einecsNumber' : 251-780-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only