missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Asp-Glu-Val-Asp-al

Product Code. 15406789
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reversible inhibitor of interleukin-1beta converting enzyme (ICE); also inhibits caspase-3 cleavage of poly(ADP-ribose) polymerase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169332-60-9
Molecular Formula C20H30N4O11
Synonym Ac-Asp-Glu-Val-Asp-CHO; Ac-DEVD-CHO

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Reversible inhibitor of interleukin-1beta converting enzyme (ICE); also inhibits caspase-3 cleavage of poly(ADP-ribose) polymerase
Formula Weight 502.47
Sensitivity Moisture Sensitive
Product Type N-Acetyl-Asp-Glu-Val-Asp-al
Form Powder
InChI Key UMBVAPCONCILTL-MRHIQRDNSA-N
SMILES CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C=O
IUPAC Name (4S)-4-[(2S)-3-carboxy-2-acetamidopropanamido]-4-{[(1S)-1-{[(2S)-1-carboxy-3-oxopropan-2-yl]carbamoyl}-2-methylpropyl]carbamoyl}butanoic acid
Molecular Weight (g/mol) 502.48
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'