missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-Asp-Glu-Val-Asp-7-amino-4-(trifluoromethyl)coumarin
Click to view available options
Quantity:
10 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 10 mg |
Description | White |
Content And Storage | -30°C to -10°C |
For Use With (Application) | A fluorogenic substrate for caspase-3 |
CAS | 201608-14-2 |
Molecular Formula | C30H34F3N5O13 |
Formula Weight | 729.61g/mol |
Synonym | Ac-DEVD-AFC |
Product Type | Signal Transduction Reagent |
Form | Powder |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'