missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine, 98%, Thermo Scientific™

Product Code. 15422109
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150812-21-8
Molecular Formula C13H12FN3O2
Molecular Weight (g/mol) 261.256
MDL Number MFCD12828054
InChI Key XTDZJOIEYRRRGJ-UHFFFAOYSA-N
Synonym n1-4-fluorobenzyl-3-nitrobenzene-1,4-diamine, 1-n-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine, 2-amino-5-4-fluorobenzyl amino-1-nitrobenzene, fluorophenylmethylnitrobenzenediamine, n1-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine, 4-4-fluorobenzylamino-2-nitroaniline, n-4-fluorobenzyl-3-nitro-p-phenylenediamine, n1-4-fluorobenzyl-3-nitro-1,4-phenylenediamine, n∼4∼-4-fluorophenyl methyl-2-nitrobenzene-1,4-diamine, n4-4-fluorophenyl methyl-2-nitro-benzene-1,4-diamine
PubChem CID 20096587
IUPAC Name 4-N-[(4-fluorophenyl)methyl]-2-nitrobenzene-1,4-diamine
SMILES C1=CC(=CC=C1CNC2=CC(=C(C=C2)N)[N+](=O)[O-])F

Specifications

Color Brown to Red
Quantity 5 g
Beilstein 22723917
Formula Weight 261.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'