missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals myo-Inositol, 98+%
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.
Solubility
Soluble in water. Slightly soluble in ethanol, dimethyl sulfoxide and alcohol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.
Solubility
Soluble in water. Slightly soluble in ethanol, dimethyl sulfoxide and alcohol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 224°C to 227°C |
Density | 1.75 |
Assay Percent Range | 98% |
Odor | Odorless |
Quantity | 2500 g |
Beilstein | 1907329 |
Merck Index | 14,4978 |
Solubility Information | Soluble in water. Slightly soluble in ethanol,dimethyl sulfoxide and alcohol. Insoluble in ether. |
Formula Weight | 180.16 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'