missing translation for 'onlineSavingsMsg'
Learn More

Murexide, For Complexometry, Honeywell Fluka™

Catalog Number p-7120606
29.12 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.


Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
3051-09-0
C8H3N5O6-2
265.141
MFCD00012777
MUMMBXDKKJEASE-UHFFFAOYSA-L
51381221
6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O
This item is not returnable. View return policy
3051-09-0
C8H3N5O6-2
265.141
MFCD00012777
MUMMBXDKKJEASE-UHFFFAOYSA-L
51381221
6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O

for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3831445
Formula Weight 284.19g/mol
Packaging Glass Bottle
Color Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Murexide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'