Learn More
Thermo Scientific™ Multidrop™ 384 Reagent Dispenser
Increase productivity with this reagent dispenser, offering continuous high-speed dispensing of liquids with excellent precision and robot compatibility.
£9473.00
Specifications
Product Type | Reagent Dispenser |
---|---|
Accuracy | 5µL: ±3% (typical), 20µL: ±2% (typical), 100µL: ±1% (typical) |
Channels | 8 |
Volume (Metric) | 5 to 395 μL |
Increments | 5 μL |
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
11530725
|
Thermo Scientific™
5840157 |
100/120 V | N/A | ||||||
Description
Increase productivity with Thermo Scientific™ Multidrop™ 384 Reagent Dispenser, which offers continuous high-speed dispensing of various liquids with excellent precision and robot compatibility.
The Multidrop 384 Reagent Dispenser portions a wide range of solutions into 96- and 384-well plates across a 5 to 395μL volume range, providing reliable microvolume dispensing for pharmaceutical and biotechnology laboratories.- Precise and reproducible dispensing results
- High-speed dispensing for high assay throughput
- Easy setup with quick plate and volume selection
- Easy to integrate with different robotic setups for higher throughput
- For use with standard cassettes only
Convenient, autoclavable dispensing cassette:
- Eight-channel detachable, autoclavable dispensing cassette to ensure sterile conditions and avoid cross contamination
- All reagent lines can be backflushed into the reagent bottle, minimizing the loss of expensive reagents
Includes:
One Standard tube dispensing cassette (Cat. No. 24072670)
Recommended for:
- Assay development
- Primary and secondary screening
- Compound storage
- Genomics and proteomics research
- Cell based assays
- ELISA assays
Specifications
Reagent Dispenser | |
8 | |
5 μL | |
96-, 384-well plates | |
96-well plate: 20µL in 5 seconds, 50µL in 8 seconds and 384-well plate: 20µL in 20 seconds, 50µL in 25 seconds | |
12.2 in. | |
6.1 in. | |
12.6 in. | |
13.7 lb. | |
50/60 Hz |
5µL: ±3% (typical), 20µL: ±2% (typical), 100µL: ±1% (typical) | |
5 to 395 μL | |
RS-232 | |
5μL: CV≤10% (typical), 20μL: CV≤1.5% (typical), | |
<7 mL (reagent recovery option) | |
31 cm | |
15.5 cm | |
32 cm | |
6.2 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.