missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ mPES MicroKos™ Filter Modules, Sterile
Spectrum Labs™ mPES MicroKos™ Filter Modules, Sterile are single-use, hollow fiber filters designed for tangential flow filtration.
£143.00 - £219.00
Specifications
Material | Modified PES |
---|---|
Sterility | Sterile |
Type | Hollow Fiber Filtration Module |
Product Code | Brand | Diameter (Metric) Inside | Includes | Length (Metric) | MWCO | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Includes | Length (Metric) | MWCO | Price | Quantity & Availability | ||
12447051
|
Spectrum Labs™
C02-E003-05-S |
Fiber ID: 0.5mm, Housing ID: 3.2mm, Inlet/Retentate ID: 3mm, Side Port ID: 4.3mm | Inlet/Outlet Ports: Male Luer-Lok, Side Port: Female Luer-Lok | Total Length: 23cm, Effective length: 20cm | 3 kDa |
£143.00
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||
12447061
|
Spectrum Labs™
C02-E100-05-S |
Fiber ID: 0.5mm, Housing ID: 3.1mm, Inlet/Retentate ID: 3mm, Side Port ID: 4.3mm | Inlet/Outlet Ports: Male Luer-Lok, Side Port: Female Luer-Lok | Total Length: 23cm, Effective length: 20cm | 100 kDa |
£219.00
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
Description
- Designed for tangential flow filtration of small volumes ranging from 1mL to 100mL
- Single-use MicroKros™ hollow fiber filters are the first efficient and practical tangential flow filtration device for characterizing a process for scale-up or filtration of small volumes in a laboratory setting.
- MicroKros™ are available with MLL x FLL connections and 1 Effective Length
Specifications
Modified PES | |
Sterile | |
Hollow Fiber Filtration Module |