Learn More
MP Biomedicals™ Protease Inhibitor Cocktail Kit
Research Use Only
Brand: MP Biomedicals™ 0215883701
Code : NEW
Additional Details : Weight : 0.10000kg Transport : UN number : 3261 Chem class : 8 Pack group : II
Description
This convenient kit contains the following protease inhibitors in pre-weighed vials, to prepare 100ml of inhibitor cocktail:
- AEBSF: 40mg
- EDTA-Na2: 500mg
- Leupeptin: 0.1mg
- Pepstatin A: 0.1mg
Specifications
AEBSF Hydrochloride, EDTA Disodium Dihydrate, Leupeptin Hemisulfate, Pepstatin A | |
2°C to 8°C | |
Each |
Protease Inhibitor Cocktail | |
Protease inhibito r |
Safety and Handling
- Protease Inhibitor Cocktail
Signal Word
- Danger
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with all local, regional,national, and international regulations..