missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM, Powdered Media

Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media.

Brand:  MP Biomedicals™ 114500022

531.70 GBP valid until 2024-05-31
Use promo code "21654" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11327889

  • £559.68 / 100g
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

Formulation

  • Add 0.79g of CSM to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae

Applications:

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Specifications

Molecular biology Certified™
Powder
Yeast Media
For growth and maintenance of M13 Phage or other filamentous Phage
15°C to 30°C
Molecular biology
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, Uracil 20mg/L, L-Tyrosine 50mg/L, L-Valine 140mg/L, Total 790mg/L
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers