missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-HIS-LEU-TRP-URA, Powdered Media
Dropout supplement for S. cereviseae
£79.29 - £634.94
Specifications
Certifications/Compliance | Molecular biology Certified™ |
---|---|
Content And Storage | 15°C to 30°C |
Form | Powder |
Product Type | Yeast Media |
Formulation | Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307939
|
MP Biomedicals™
114540012 |
10 g |
£79.29
10g |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11317939
|
MP Biomedicals™
114540022 |
100 g |
£634.94
100g |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
- Four dropout
- Supplement mixture of amino acids
- Complete formulations
- Dropout formulations
Formulation
- Add 0.60g of CSM-His-Leu-Trp-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined four dropout medium
Applications:
Yeast cell culture, Gene expression, Transcription
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |