missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2b Isotype Control (PLRV219), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA110427
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10427 can be used as a negative control to measure the non-specific binding of Mouse IgG2b in human, porcine, mouse and rat tissues.
Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2b | |
PLRV219 | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Potato leaf roll virus | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG2b |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |