missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enrofloxacin, Mouse, Clone: 7F21G1F71, Abnova™
Mouse monoclonal antibody raised against Enrofloxacin.
Brand: Abnova MAB2280.100ug
This item is not returnable.
View return policy
Specifications
Enrofloxacin | |
Monoclonal | |
Unconjugated | |
ELISA (1000-1:2000) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgM |
ELISA | |
7F21G1F71 | |
Mouse monoclonal antibody raised against Enrofloxacin. | |
In PBS, pH 7.4 | |
Enrofloxacin conjugated with KLH. | |
RUO | |
This antibody reacts equally well with enrofloxacin and its main metabolite, ciprofloxacin. | |
Liquid |