Learn More
CLCA2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CLCA2.
Marke: Abnova H00009635-A01.50uL
Weitere Details : Weight : 0.00010kg
Beschreibung
The protein encoded by this gene belongs to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. Since this protein is expressed predominantly in trachea and lung, it is suggested to play a role in the complex pathogenesis of cystic fibrosis. It may also serve as adhesion molecule for lung metastatic cancer cells, mediating vascular arrest and colonization, and furthermore, it has been implicated to act as a tumor suppressor gene for breast cancer. [provided by RefSeq
Sequence: PTFSLVQAGDKVVCLVLDVSSKMAEADRLLQLQQAAEFYLMQIVEIHTFVGIASFDSKGEIRAQLHQINSNDDRKLLVSYLPTTVSAKTDISICSGLKKGFSpezifikation
CLCA2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CLCA2. | |
CLCA2 | |
CACC/CACC3/CLCRG2/CaCC-3/FLJ97885 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006536 | |
CLCA2 | |
CLCA2 (NP_006527, 300 a.a. to 400 a.a) partial recombinant protein with GST tag. | |
RUO | |
9635 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only