missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYPRO™ Protein Gel Stain Starter Kit

Makes it easy for first-time users to try three SDS-PAGE stains

Brand:  Invitrogen™ S12012

Code : 50

Additional Details : Weight : 0.00450kg

Product Code. 10462872

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: 50μL of SYPRO Orange dye concentrate (S-6650), 50μL of SYPRO Red dye concentrate (S-6653), 50μL of SYPRO Tangerine dye concentrate (S-12010) and a SYPRO protein gel stain photographic filter (S-6656).

Description

Description

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature and protect from light.
Fluorescent
Protein Gel Stain Kit
Room Temperature
In-Gel Detection
SYPRO Tangerine, SYPRO Orange, SYPRO Red
SYPRO™
Protein
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.