Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
Brand: Epredia™ 88024
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
- Background is stained green
Specifications
Fast Green Stain Solution | |
125 mL |