Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
Brand: Epredia™ 88023
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
- Background is stained green
Specifications
Methenamine/Borax | |
6 Capsules |
Safety and Handling
- Danger
Hazard Category
- Flammable solid Category 2
- Skin sensitiser Category 1
- Reproductive toxicity Category 1B
Hazard Statement
- H228-Flammable solid.
- H317-May cause an allergic skin reaction.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : borates tetra sodiumsalts decahydrate,methenamine