Learn More
Hamilton™ 1000 Series Standard PTFE Instrument Syringes without Needles
Setting the standard for quality and reliability.
Brand: Hamilton™ 9880-40
Code : NEW
Additional Details : Weight : 0.06300kg
Description
- Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
- Enables precision liquid handling
- Gastight for leak-free performance
Specifications
2.5 mL |