Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood and Face Seal
Product Code. 15516464
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Fits full-face respirators and creates a close seal. Ansell Microchem™ 4000 Coveralls with Hood and Face Seal protect against dangerously concentrated chemicals and biological agents.
  • Coverall with elasticated 2-piece hood with neoprene rubber face seal
  • Attached Ansell Barrier Gloves, with oversleeves and finger loops
  • Integrated socks and boot overflap
  • Rear horizontal zip entry
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size 2X-Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Waist Style Elastic
Ankle Style Elastic, Attached Socks, Boot Overflaps
Cuff Style Double, Elastic, Attached Gloves
Neck Style Hooded
Hood Type Attached, Neoprene Rubber Face Seal
Closure Type Rear-entry Horizontal Zipper
Seam Type Taped, Welded
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
Description Hood with neoprene face seal, attached Ansell Barrier™ Gloves, integrated socks with boot overflap
No. of Pockets None
Style Includes Hood with neoprene face seal, attached Ansell Barrier™ Gloves, integrated socks with boot overflap