Learn More
METTL24 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557952
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AQSLDEEAWR FLRYISTTQI ACNHMNTDSL ATDSSPTHKP WSVCLDDRFN LAHQIRNKQC RLYSLGLGSD DTHFEVSMAN NGCEV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 80%.
METTL24 gene ontology annotations related to this gene include methyltransferase activity.Specifications
METTL24 | |
Polyclonal | |
Unconjugated | |
METTL24 | |
9030224M15Rik; C6orf186; C9H6orf186; dJ71D21.2; methyltransferase like 24; methyltransferase-like protein 24; Mettl24; UPF0624 protein C6orf186; UPF0624 protein C6orf186 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
728464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JXM2 | |
METTL24 | |
Recombinant protein corresponding to Human METTL24 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only