Learn More
FAM119A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557026
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EHLCSNHSVI LLACRIRYER DNNFLAMLER QFIVRKVHYD PEKDVHIYEA QKRNQKEDL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
FAM119A is a protein-lysine methyltransferase that selectively trimethylates residues in heat shock protein 70 (HSP70) family members. Contributes to the in vivo trimethylation of Lys residues in HSPA1 and HSPA8. In vitro methylates Lys-561 in HSPA1, Lys-564 in HSPA2, Lys-585 in HSPA5, Lys-563 in HSPA6 and Lys-561 in HSPA8.Specifications
FAM119A | |
Polyclonal | |
Unconjugated | |
METTL21A | |
2310038H17Rik; AI464204; Fam119a; family with sequence similarity 119, member A; HCA557b; heat shock protein 70kDa lysine (K) methyltransferase; hepatocellular carcinoma-associated antigen 557b; HSPA lysine methyltransferase; HSPA-KMT; LOC102553119; methyltransferase like 21A; methyltransferase like 21A; protein N-lysine methyltransferase METTL21A; methyltransferase-like protein 21A; methyltransferase-like protein 21A; protein N-lysine methyltransferase METTL21A; Mettl21A; Protein N-lysine methyltransferase METTL21A; protein N-lysine methyltransferase METTL21A-like; protein-lysine methyltransferase METTL21A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXB1 | |
METTL21A | |
Recombinant protein corresponding to Human METTL21A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only