Learn More
METT10D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554185
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DERSEEKGGV EVLESCQGSS NGAQDQEASE QFGSPVAERG KRLPGVAGQY LFKCLINVKK EVDDALVEMH WVE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
METT10D is a probable methyltransferase.Specifications
METT10D | |
Polyclonal | |
Unconjugated | |
METTL16 | |
2610100D03Rik; 2810013M15Rik; A830095F14Rik; AI846133; methyltransferase 10 domain containing; methyltransferase 10 domain-containing protein; methyltransferase like 16; methyltransferase-like protein 16; METT10D; METTL16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79066 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86W50 | |
METTL16 | |
Recombinant protein corresponding to Human METTL16 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only