missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 18.4°C |
Color | Colorless |
Density | 1.1000g/mL |
Boiling Point | 189.0°C |
Quantity | 500 mL |
Flash Point | 87°C |
Packaging | Glass Bottle |
Physical Form | Liquid at 20°C |
Refractive Index | 1.4770 to 1.4800 |
Assay Percent Range | 99.8+% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'