missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange solution, Honeywell Fluka™

Product Code. 15664290
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, 0.1%, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

UN Number UN1993
Beilstein 4732884
Formula Weight 327.33g/mol
Percent Purity 0.10%
Quantity 1 L
Flash Point 39°C (102.2°F)
Chemical Name or Material Methyl Orange solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'