Learn More
Methanol, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep S-2380
Code : 04
Additional Details : Weight : 0.01000kg
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Methanol | 67-56-1 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Methanol | |
2mL amber prescored glass ampule | |
99.9 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
H2O |
67-56-1,7732-18-5 | |
1 mL | |
99.9 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
MFCD00004595 | |
CO | |
32.04 | |
CHEBI:17790 |
Product intended for laboratory use only