missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric acid, Thermo Scientific Chemicals

£47.03 - £629.13

Chemical Identifiers

CAS 37267-86-0, 50813-16-6
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15169313
View Documents Promotion Details
Thermo Scientific Acros
450251000
100 g Glass Bottle
£47.03
100g
Please to purchase this item. Need a web account? Register with us today!
15179313
View Documents Promotion Details
Thermo Scientific Acros
450255000
500 g Glass Bottle
£155.78
500g
Please to purchase this item. Need a web account? Register with us today!
15159313
View Documents Promotion Details
Thermo Scientific Acros
450250025
2.5 kg Glass Bottle
£629.13
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37267-86-0, 50813-16-6
79.979
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
OP(=O)=O
HO3P
UEZVMMHDMIWARA-UHFFFAOYSA-N
3084658

Specifications

37267-86-0, 50813-16-6
(HPO3)n
Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol
OP(=O)=O
3084658
Metaphosphoric acid
HO3P
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
UEZVMMHDMIWARA-UHFFFAOYSA-N
79.979
ACS Reagent
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Metaphosphoric acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only