Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 2, 3 or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand:  Hamilton™ 7748-07
Code : A0
Additional Details : Weight : 1.00000kg
Product Code. 11522345
  • £166.00 / Pack of 6
Estimated Shipment: 21-06-2024
to see stock.
Description
Description
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Specifications
Point 2, 3 or 4