missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Mesothelin Ab-1 Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelin Ab-1, Mouse Monoclonal Antibody.
£98.12 - £625.00
Specifications
Antigen | Mesothelin Ab-1 |
---|---|
Clone | 5B2 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12689556
|
Epredia™
MS-1320-S1 |
Concentrated | 500 μL |
£323.00
500µL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649556
|
Epredia™
MS-1320-S |
Concentrated | 1 mL |
£625.00
1mL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12679556
|
Epredia™
MS-1320-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659556
|
Epredia™
MS-1320-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, that is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.Host Species: Mouse
Clone: 5B2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule
Molecular Weight: 40kDa
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mesothelin Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG1 |
5B2 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule | |
Primary |