Learn More
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 7.5
£110.55 - £150.75
Chemical Identifiers
CAS | 145224-94-8 |
---|---|
Molecular Formula | C6H15NO5S |
Molecular Weight (g/mol) | 213.248 |
MDL Number | MFCD00283848 |
InChI Key | MIIIXQJBDGSIKL-UHFFFAOYSA-N |
PubChem CID | 16218417 |
IUPAC Name | 2-morpholin-4-ylethanesulfonic acid;hydrate |
SMILES | C1COCCN1CCS(=O)(=O)O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485059
|
Thermo Scientific Chemicals
J62752.AK |
250 mL |
£110.55
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15495059
|
Thermo Scientific Chemicals
J62752.AP |
500 mL |
£150.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.
Solubility
Soluble in water (0.5 M at 20°C).
Notes
Solutions are stable at 2-8°C for months. Sterilize by filtration through 0.2 mm filters. Autoclaving is not recommended for any sulfonic acid buffer. If buffers must be nuclease-free, treat the water first, then add the buffer after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably). The identity of the yellow breakdown product is unknown.
Chemical Identifiers
145224-94-8 | |
213.248 | |
MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid;hydrate |
C6H15NO5S | |
MFCD00283848 | |
16218417 | |
C1COCCN1CCS(=O)(=O)O.O |
Specifications
145224-94-8 | |
7.5 | |
C6H15NO5S | |
Soluble in water (0.5 M at 20°C). | |
C1COCCN1CCS(=O)(=O)O.O | |
213.248 | |
MES buffer soln. |
1.0 M | |
Liquid | |
MFCD00283848 | |
MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid;hydrate | |
16218417 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only