missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Iodide Red, purum p.a, ≥99.0% (RT), Honeywell Fluka™

Catalog Number p-7120558
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7774-29-0
HgI2
454.401
MFCD00011044
YFDLHELOZYVNJE-UHFFFAOYSA-L
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
24485
CHEBI:49659
diiodomercury
I[Hg]I
This item is not returnable. View return policy
7774-29-0
HgI2
454.401
MFCD00011044
YFDLHELOZYVNJE-UHFFFAOYSA-L
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
24485
CHEBI:49659
diiodomercury
I[Hg]I

Puriss. p.a., ACS Reagent, red,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 254°C
Melting Point 259°C
Color Red
Physical Form Powder
Quantity 10 g
UN Number UN1638
Formula Weight 454.4g/mol
Percent Purity ≥99.0% (RT)
Odor Odorless
Packaging Glass bottle
Chemical Name or Material Mercury(II) iodide red
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'